• USA Home
  • 8.43724 - Tetramethylammonium iodide

EMAIL THIS PAGE TO A FRIEND
8.43724 EMD Millipore

Tetramethylammonium iodide

Tetramethylammonium iodide for synthesis. CAS 75-58-1, molar mass 201.05 g/mol., for synthesis

  • CAS Number 75-58-1

  • Linear Formula C₄H₁₂IN

  • Molecular Weight 201.05 g/mol

  •  EC Number 200-881-3

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 g in Glass bottle (8437240025)
density   1.84 g/cm3
mp   >300 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (argentometric): ≥ 99.0 %
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2923 90 00
RTECS 
PA1050000
Storage class 
6.1A Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials
WGK 
WGK 3 highly water endangering
Declaration (railroad and road) ADR, RID 
UN 2811 , 6.1, II
Declaration (transport by air) IATA-DGR 
UN 2811 , 6.1, II
Declaration (transport by sea) IMDG-Code 
UN 2811 , 6.1, II
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?