• USA Home
  • 8.43779 - Tris(trimethylsilyl)silane

EMAIL THIS PAGE TO A FRIEND
8.43779 EMD Millipore

Tris(trimethylsilyl)silane

Tris(trimethylsilyl)silane for synthesis. CAS 1873-77-4, molar mass 248.67 g/mol., for synthesis

  • CAS Number 1873-77-4

  • Linear Formula C₉H₂₈Si₄

  • Molecular Weight 248.67 g/mol

Purchase

Properties

packaging   5 ml in Glass bottle (8437790005)
bp   82 - 84 °C (16 hPa)
density   0.806 g/cm3
flash point   55 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 96.0 % (a/a)
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2931 90 00
Hazard Statement(s) 
H226: Flammable liquid and vapour.
Precautionary Statement(s) 
P210: Keep away from heat.
Storage class 
3 Flammable liquids
WGK 
WGK 1 slightly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS02  GHS02
Declaration (railroad and road) ADR, RID 
UN 1993 , 3, II
Declaration (transport by air) IATA-DGR 
UN 1993 , 3, II
Declaration (transport by sea) IMDG-Code 
UN 1993 , 3, II
Categories of danger 
flammable
R Phrase 
R 10
Flammable.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?