EMAIL THIS PAGE TO A FRIEND
8.43783 EMD Millipore

γ-Nonanolactone

γ-Nonanolactone for synthesis. CAS 104-61-0, molar mass 156.23 g/mol., for synthesis

  • CAS Number 104-61-0

  • Linear Formula C₉H₁₆O₂

  • Molecular Weight 156.23 g/mol

  •  EC Number 203-219-1

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   500 ml in Glass bottle (8437830500)
bp   121 - 122 °C (8 hPa)
density   0.97 g/cm3 (20 °C)
flash point   113 °C
LD 50 oral   LD50 Rat 6600 mg/kg
LD 50 dermal   LD50 Rabbit > 5000 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Density (d 20 °C/ 4 °C): 0.960 - 0.965
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2932 20 90
RTECS 
LU3675000
Storage class 
10 Combustible liquids not in Storage Class 3
WGK 
WGK 1 slightly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?