• USA Home
  • 8.43787 - 4,4-Dimethyl-2-cyclohexen-1-one

EMAIL THIS PAGE TO A FRIEND
8.43787 EMD Millipore

4,4-Dimethyl-2-cyclohexen-1-one

4,4-Dimethyl-2-cyclohexen-1-one for synthesis. CAS 1073-13-8, molar mass 124.18 g/mol., for synthesis

  • CAS Number 1073-13-8

  • Linear Formula C₈H₁₂O

  • Molecular Weight 124.18 g/mol

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   5 ml in Glass bottle (8437870005)
bp   72 - 73 °C (27 hPa)
density   0.930 - 0.935 g/cm3 (20 °C)
flash point   64 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 97.0 % (a/a)
  Density (d 20 °C/ 4 °C): 0.930 - 0.935
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2914 29 00
Storage class 
10 Combustible liquids not in Storage Class 3
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?