• USA Home
  • 8.43800 - 3-(2-Ethylhexyloxy)propylamine

EMAIL THIS PAGE TO A FRIEND
8.43800 EMD Millipore

3-(2-Ethylhexyloxy)propylamine

3-(2-Ethylhexyloxy)propylamine for synthesis. CAS 5397-31-9, molar mass 187.33 g/mol., for synthesis

  • CAS Number 5397-31-9

  • Linear Formula C₁₁H₂₅NO

  • Molecular Weight 187.33 g/mol

  •  EC Number 226-420-6

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 ml in Glass bottle (8438000025)
bp   234 °C (1013 hPa)
expl. lim.     1 - 4.7 %(V)
vapor pressure   <1 hPa (20 °C)
solubility   15 g/l
density   0.848 g/cm3 (20 °C)
flash point   103 °C
autoignition temp.   220 °C
mp   -90 °C
LD 50 oral   LD50 Rat 320 mg/kg
LD 50 dermal   LD50 Rabbit 305 mg/kg
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 99.0 % (a/a)
  Density (d 20 °C/ 4 °C): 0.845 - 0.849
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2922 19 00
Hazard Statement(s) 
H302: Harmful if swallowed.
H311: Toxic in contact with skin.
H314: Causes severe skin burns and eye damage.
H411: Toxic to aquatic life with long lasting effects.
Precautionary Statement(s) 
P273: Avoid release to the environment.
P280: Wear protective gloves/ protective clothing/ eye protection/ face protection.
P301 + P330 + P331: IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
P308 + P310: IF exposed or concerned: immediately call a POISON CENTER or doctor/ physician.
RTECS 
UI2820000
Storage class 
6.1A Combustible substances, toxic
WGK 
WGK 3 highly water endangering
Hazard Pictogram(s) 
GHS05  GHS05
Hazard Pictogram(s) 
GHS06  GHS06
Hazard Pictogram(s) 
GHS09  GHS09
Hazard Symbols 
Corrosive  Corrosive
Hazard Symbols 
Dangerous for the environment  Dangerous for the environment
Hazard Symbols 
Toxic  Toxic
Declaration (railroad and road) ADR, RID 
UN 2922 , 8 (6.1), III
Declaration (transport by air) IATA-DGR 
UN 2922 , 8 (6.1), III
Declaration (transport by sea) IMDG-Code 
UN 2922 , 8 (6.1), III, Segregation Group: 18 (Alkalis)
Categories of danger 
toxic, corrosive
R Phrase 
R 22-24-34
Harmful if swallowed.Toxic in contact with skin.Causes burns.
S Phrase 
S 26-36/37/39-45-61
In case of contact with eyes, rinse immediately with plenty of water and seek medical advice.Wear suitable protective clothing, gloves and eye/face protection.In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).Avoid release to the environment. Refer to special instructions/ Safety data sheets.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?