• USA Home
  • 8.43802 - 2,5-Dimethyl-4-hydroxy-(2H)-furan-3-on

EMAIL THIS PAGE TO A FRIEND
8.43802 EMD Millipore

2,5-Dimethyl-4-hydroxy-(2H)-furan-3-on

2,5-Dimethyl-4-hydroxy-(2H)-furan-3-on for synthesis. CAS 3658-77-3, molar mass 128.13 g/mol., for synthesis

  • CAS Number 3658-77-3

  • Linear Formula C₆H₈O₃

  • Molecular Weight 128.13 g/mol

  •  EC Number 222-908-8

Purchase

Properties

Related Categories Biochemicals and Reagents More...
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2932 19 00
RTECS 
LU3990000
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?