• USA Home
  • 8.43804 - Tetrabutylammonium bromide

EMAIL THIS PAGE TO A FRIEND
8.43804 EMD Millipore

Tetrabutylammonium bromide

(50% solution in water) for synthesis

Purchase

Properties

packaging   100 ml in Glass bottle (8438040100)
density   1.043 g/cm3
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (argentometric): 47.0 - 53.0 %

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2923 90 00
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?