• USA Home
  • 8.43806 - Bis(1,5-cyclooctadiene)nickel(0)

EMAIL THIS PAGE TO A FRIEND
8.43806 EMD Millipore

Bis(1,5-cyclooctadiene)nickel(0)

Bis(1,5-cyclooctadiene)nickel(0) for synthesis. CAS No. 1295-35-8, EC Number 215-072-0., for synthesis

  • CAS Number 1295-35-8

  • Linear Formula C₁₆H₂₄Ni

  • Molecular Weight 275.08 g/mol

  •  EC Number 215-072-0

Purchase

Properties

packaging   1 g in Glass bottle (8438060001)
mp   60 °C (decomposition)
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (of Ni): ≥ 96.0 %
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2931 90 00
Hazard Statement(s) 
H228: Flammable solid.
H317: May cause an allergic skin reaction.
Precautionary Statement(s) 
P210: Keep away from heat/sparks/open flames/hot surfaces. No smoking.
P262: Do not get in eyes, on skin, or on clothing.
P280: Wear protective gloves.
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
RTECS 
QR6135000
Storage class 
4.1B Flammable solid hazardous materials
WGK 
WGK 3 highly water endangering
Disposal 
15
Heavy metal-containing solutions and solids: container E. Stir Raney nickel (also: Urushibara nickel) in the form of an aqueous suspension into hydrochloric acid (Cat. No. 100312) until dissolved (container E). Neither Raney nickel itself nor any filter residues should be allowed to dry out, otherwise they will spontaneously ignite in air. In this context, heavy metal means any compound of antimony, arsenic, cadmium, chromium(VI), copper, lead, nickel and tin, as well as these subtances in metallic form, if they are classified as hazardous (acc. to AbfallverzeichnisV - Waste Catalogue Ordinance, Appendix 3). Other heavy metals should be collected separately.
Hazard Pictogram(s) 
GHS02  GHS02
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Flammable  Flammable
Hazard Symbols 
Irritant  Irritant
Declaration (railroad and road) ADR, RID 
UN 1325 , 4.1, II
Declaration (transport by air) IATA-DGR 
UN 1325 , 4.1, II
Declaration (transport by sea) IMDG-Code 
UN 1325 , 4.1, II
Categories of danger 
highly flammable, sensitizing
R Phrase 
R 11-43
Highly flammable.May cause sensitisation by skin contact.
S Phrase 
S 16-24-37
Keep away from sources of ignition - No smoking.Avoid contact with skin.Wear suitable gloves.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?