• USA Home
  • 8.43812 - Hexachloroplatinic(IV) acid (11-15% Pt),

EMAIL THIS PAGE TO A FRIEND
8.43812 EMD Millipore

Hexachloroplatinic(IV) acid (11-15% Pt),

25-50% solution in isopropanol, for synthesis

Purchase

Properties

packaging   1 ml in Glass bottle (8438120001)
bp   82.4 °C (1013 hPa) (2-Propanol)
expl. lim.     2 - 12.7 %(V) (2-Propanol)
vapor pressure   43 hPa (20 °C)
density   1.033 g/cm3 (20 °C)
flash point   12 °C (2-Propanol)
autoignition temp.   425 °C (2-Propanol)
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (of Pt): 11.0 - 15.0 %

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
3824 99 92
Hazard Statement(s) 
H225: Highly flammable liquid and vapour.
H301: Toxic if swallowed.
H314: Causes severe skin burns and eye damage.
H317: May cause an allergic skin reaction.
H334: May cause allergy or asthma symptoms or breathing difficulties if inhaled.
H336: May cause drowsiness or dizziness.
Precautionary Statement(s) 
P210: Keep away from heat/sparks/open flames/hot surfaces. No smoking.
P240: Ground/bond container and receiving equipment.
P280: Wear protective gloves/ protective clothing/ eye protection/ face protection.
P301 + P330 + P331: IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P304 + P340: IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
P342 + P311: If experiencing respiratory symptoms: Call a POISON CENTER/doctor.
P308 + P310: IF exposed or concerned: immediately call a POISON CENTER or doctor/ physician.
P403 + P233: Store in a well-ventilated place. Keep container tightly closed.
Storage class 
3 Flammable Liquids
WGK 
WGK 3 highly water endangering
Disposal 
27
Residues containing valuable recoverable metals should be forwarded for recycling. Container H.
Hazard Pictogram(s) 
GHS02  GHS02
Hazard Pictogram(s) 
GHS05  GHS05
Hazard Pictogram(s) 
GHS06  GHS06
Hazard Pictogram(s) 
GHS08  GHS08
Hazard Symbols 
Flammable  Flammable
Hazard Symbols 
Toxic  Toxic
Declaration (railroad and road) ADR, RID 
UN 3286 , 3 (6.1, 8), II
Declaration (transport by air) IATA-DGR 
UN 3286 , 3 (6.1, 8), II
Declaration (transport by sea) IMDG-Code 
UN 3286 , 3 (6.1, 8), II, Segregation Group: 1 (Acids)
Categories of danger 
highly flammable, toxic, corrosive, sensitizing
R Phrase 
R 11-25-34-42/43-67
Highly flammable.Toxic if swallowed.Causes burns.May cause sensitisation by inhalation and skin contact.Vapours may cause drowsiness and dizziness.
S Phrase 
S 16-23-26-36/37/39-45
Keep away from sources of ignition - No smoking.Do not breathe vapour.In case of contact with eyes, rinse immediately with plenty of water and seek medical advice.Wear suitable protective clothing, gloves and eye/face protection.In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?