• USA Home
  • 8.43813 - 3-Methoxy-1-propanol

EMAIL THIS PAGE TO A FRIEND
8.43813 EMD Millipore

3-Methoxy-1-propanol

3-Methoxy-1-propanol for synthesis. CAS 1589-49-7, molar mass 90.12 g/mol., for synthesis

  • CAS Number 1589-49-7

  • Linear Formula C₄H₁₀O₂

  • Molecular Weight 90.12 g/mol

  •  EC Number 433-230-9

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   10 ml in Glass bottle (8438130010)
bp   149 - 150 °C (1013 hPa)
density   0.94 g/cm3 (20 °C)
flash point   38 °C
LD 50 oral   LD50 Rat 5710 mg/kg
LD 50 dermal   LD50 Rabbit 5320 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Density (d 20 °C/ 4 °C): 0.916 - 0.920
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2909 49 80
Hazard Statement(s) 
H226: Flammable liquid and vapour.
Precautionary Statement(s) 
P210: Keep away from heat.
P262: Do not get in eyes, on skin, or on clothing.
RTECS 
UB7650000
Storage class 
3 Flammable liquids
WGK 
WGK 1 slightly water endangering
Disposal 
1
Strongly contaminated halogen-free organic solvents: container A.
Hazard Pictogram(s) 
GHS02  GHS02
Declaration (railroad and road) ADR, RID 
UN 1987 , 3, III
Declaration (transport by air) IATA-DGR 
UN 1987 , 3, III
Declaration (transport by sea) IMDG-Code 
UN 1987 , 3, III
Categories of danger 
flammable
R Phrase 
R 10
Flammable.
S Phrase 
S 24
Avoid contact with skin.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?