• USA Home
  • 8.43815 - 1,3,5-Triazine-2,4,6(1H,3H,5H)-thrithione

EMAIL THIS PAGE TO A FRIEND
8.43815 EMD Millipore

1,3,5-Triazine-2,4,6(1H,3H,5H)-thrithione

1,3,5-Triazine-2,4,6(1H,3H,5H)-thrithione for synthesis. CAS 638-16-4, molar mass 177.27 g/mol., for synthesis

  • CAS Number 638-16-4

  • Linear Formula C₃H₃N₃S₃

  • Molecular Weight 177.27 g/mol

  •  EC Number 211-322-8

Purchase

Properties

Related Categories Biochemicals and Reagents, Building Blocks, Chemical Synthesis, Heterocyclic Building Blocks, Thiazines More...
packaging   25 g in Glass bottle (8438150025)
mp   300 °C
LD 50 oral   LD50 Rat 9500 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (acidimetric): ≥ 97.0 %
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2933 69 80
RTECS 
XZ2830000
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?