• USA Home
  • 8.43817 - N-Acetylglycine ethyl ester

EMAIL THIS PAGE TO A FRIEND
8.43817 EMD Millipore

N-Acetylglycine ethyl ester

N-Acetylglycine ethyl ester for synthesis. CAS 1906-82-7, EC Number 217-608-9, chemical formula CH₃-C(O)NH-CH₂-C(O)O-CH₂-CH₃., for synthesis

  • CAS Number 1906-82-7

  • Linear Formula C₆H₁₁NO₃

  • Molecular Weight 145.16 g/mol

  •  EC Number 217-608-9

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 g in Glass bottle (8438170025)
bp   260 °C (950 hPa)
flash point   113 °C
mp   43 - 46 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 99.0 % (a/a)
  Melting range (lower value): ≥ 45 °C
  Melting range (upper value): ≤ 48 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2924 19 00
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 1 slightly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?