• USA Home
  • 8.43819 - 1,3-Dicyclohexylurea

EMAIL THIS PAGE TO A FRIEND
8.43819 EMD Millipore

1,3-Dicyclohexylurea

1,3-Dicyclohexylurea for synthesis. CAS 2387-23-7, molar mass 224.35 g/mol., for synthesis

  • CAS Number 2387-23-7

  • Linear Formula C₁₃H₂₄N₂O

  • Molecular Weight 224.35 g/mol

  •  EC Number 219-213-7

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 g in Plastic bottle (8438190025)
density   1.15 g/cm3 (20 °C)
mp   230 - 234 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (HClO₄): ≥ 98.0 %
  Melting range (lower value): ≥ 230 °C
  Melting range (upper value): ≤ 234 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2924 21 00
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 1 slightly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?