• USA Home
  • 8.43822 - Bis(trifluoromethylsulfonyl)imide

EMAIL THIS PAGE TO A FRIEND
8.43822 EMD Millipore

Bis(trifluoromethylsulfonyl)imide

Bis(trifluoromethylsulfonyl)imide for synthesis. CAS 82113-65-3, pH 1 (500 g/l, H₂O, 20 °C)., for synthesis

  • CAS Number 82113-65-3

  • Linear Formula C₂HF₆NO₄S₂

  • Molecular Weight 281.15 g/mol

  •  EC Number 435-300-4

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   5 g in Glass bottle (8438220005)
solubility   800 g/l Risk of violent reaction.
density   2.15 g/cm3 (18.2 °C)
mp   46 - 53 °C
pH   0.8 (100 g/l, H₂O, 20 °C) Risk of violent reaction.
LD 50 oral   LD50 Rat > 50 - 200 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (NMR): ≥ 95.0 %

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2935 90 90
Hazard Statement(s) 
H301: Toxic if swallowed.
H318: Causes serious eye damage.
H412: Harmful to aquatic life with long lasting effects.
Precautionary Statement(s) 
P273: Avoid release to the environment.
P280: Wear eye protection.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
P308 + P310: IF exposed or concerned: immediately call a POISON CENTER or doctor/ physician.
Storage class 
6.1C Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK 
WGK 2 water endangering
Disposal 
9
Carcinogenic compounds and flammable compounds labelled "Highly toxic" or "Toxic": container F. Alkyl sulfates are carcinogenic; Take particular care to avoid inhalation and skin contact. To neutralize alkyl sulfates, add dropwise (from a dropping funnel) to concentrated ice-cool Ammonia solution (Cat.No. 105426) with vigorous stirring. Before placing in container D, check the pH with pH Universal indicator strips (Cat.No. 109535).
Hazard Pictogram(s) 
GHS05  GHS05
Hazard Pictogram(s) 
GHS06  GHS06
Hazard Symbols 
Toxic  Toxic
Declaration (railroad and road) ADR, RID 
UN 2811 , 6.1, III
Declaration (transport by air) IATA-DGR 
UN 2811 , 6.1, III
Declaration (transport by sea) IMDG-Code 
UN 2811 , 6.1, III
Categories of danger 
toxic, irritant, dangerous for the environment
R Phrase 
R 25-41-52/53
Toxic if swallowed.Risk of serious damage to eyes.Harmful to aquatic organisms, may cause long-term adverse effects in the aquatic environment.
S Phrase 
S 26-37/39-45-61
In case of contact with eyes, rinse immediately with plenty of water and seek medical advice.Wear suitable gloves and eye/face protection.In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).Avoid release to the environment. Refer to special instructions/ Safety data sheets.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?