• USA Home
  • 8.43823 - 2-Bromomethyl-1,3-dioxolane

EMAIL THIS PAGE TO A FRIEND
8.43823 EMD Millipore

2-Bromomethyl-1,3-dioxolane

2-Bromomethyl-1,3-dioxolane for synthesis. CAS 4360-63-8, molar mass 167 g/mol., for synthesis

  • CAS Number 4360-63-8

  • Linear Formula C₄H₇BrO₂

  • Molecular Weight 167 g/mol

  •  EC Number 224-443-6

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 ml in Glass bottle (8438230025)
bp   80 - 82 °C (36 hPa)
density   1.628 g/cm3 (20 °C)
flash point   62 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 96.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.618 - 1.622
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2932 99 00
Hazard Statement(s) 
H315: Causes skin irritation.
H319: Causes serious eye irritation.
Precautionary Statement(s) 
P260: Do not breathe vapour.
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Irritant  Irritant
Categories of danger 
irritant
R Phrase 
R 36/38
Irritating to eyes and skin.
S Phrase 
S 23
Do not breathe vapour.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?