• USA Home
  • 8.43829 - 1-Methylbenzimidazole

EMAIL THIS PAGE TO A FRIEND
8.43829 EMD Millipore

1-Methylbenzimidazole

1-Methylbenzimidazole for synthesis. CAS 1632-83-3, molar mass 132.17 g/mol., for synthesis

  • CAS Number 1632-83-3

  • Linear Formula C₈H₈N₂

  • Molecular Weight 132.17 g/mol

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   5 g in Glass bottle (8438290005)
mp   98.1 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 97.0 % (a/a)
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2933 99 80
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?