• USA Home
  • 8.43831 - 2-Chloromethyl-1,3-dioxolane

EMAIL THIS PAGE TO A FRIEND
8.43831 EMD Millipore

2-Chloromethyl-1,3-dioxolane

2-Chloromethyl-1,3-dioxolane for synthesis. CAS 2568-30-1, molar mass 122.55 g/mol., for synthesis

  • CAS Number 2568-30-1

  • Linear Formula C₄H₇ClO₂

  • Molecular Weight 122.55 g/mol

  •  EC Number 219-908-5

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   100 ml in Glass bottle (8438310100)
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 97.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.245 - 1.249
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2932 99 00
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?