• USA Home
  • 8.43837 - Rhodium (approx.5%) on alumina

EMAIL THIS PAGE TO A FRIEND
8.43837 EMD Millipore

Rhodium (approx.5%) on alumina

for synthesis

Purchase

Properties

packaging   1 g in Glass bottle (8438370001)
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay of Rh: 4.5 - 5.5 %

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
3815 12 00
Precautionary Statement(s) 
P260: Do not breathe dust.
Storage class 
10 - 13 Other liquids and solids
WGK 
NWG not water endangering
Disposal 
27
Residues containing valuable recoverable metals should be forwarded for recycling. Container H.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?