• USA Home
  • 8.43844 - Trimethyl(methylcyclopentadienyl) platinium(IV)

EMAIL THIS PAGE TO A FRIEND
8.43844 EMD Millipore

Trimethyl(methylcyclopentadienyl) platinium(IV)

Trimethyl(methylcyclopentadienyl) platinium(IV) for synthesis. CAS 94442-22-5, chemical formula C₅H₄CH₃Pt(CH₃)₃., for synthesis

  • CAS Number 94442-22-5

  • Linear Formula C₉H₁₆Pt

  • Molecular Weight 319.32 g/mol

Purchase

Properties

packaging   1 g in Glass bottle (8438440001)
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (NMR): ≥ 98.0 %

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2843 90 90
Hazard Statement(s) 
H300 + H310: Fatal if swallowed or in contact with skin
H317: May cause an allergic skin reaction.
H400: Very toxic to aquatic life.
Precautionary Statement(s) 
P273: Avoid release to the environment.
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P308 + P310: IF exposed or concerned: immediately call a POISON CENTER or doctor/ physician.
Storage class 
6.1A Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials
WGK 
WGK 3 highly water endangering
Disposal 
27
Residues containing valuable recoverable metals should be forwarded for recycling. Container H.
Hazard Pictogram(s) 
GHS06  GHS06
Hazard Pictogram(s) 
GHS09  GHS09
Hazard Symbols 
Dangerous for the environment  Dangerous for the environment
Hazard Symbols 
Toxic  Toxic
Declaration (railroad and road) ADR, RID 
UN 3467 , 6.1, I
Declaration (transport by air) IATA-DGR 
UN 3467 , 6.1, I
Declaration (transport by sea) IMDG-Code 
UN 3467 , 6.1, I
R Phrase 
R 28-27-43-50
Very toxic if swallowed.Very toxic in contact with skin.May cause sensitisation by skin contact.Very toxic to aquatic organisms.
S Phrase 
S 36/37-45-37-24-28-60-61
Wear suitable protective clothing and gloves.In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).Wear suitable gloves.Avoid contact with skin.After contact with skin, wash immediately with plenty of water.This material and its container must be disposed of as hazardous waste.Avoid release to the environment. Refer to special instructions/ Safety data sheets.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?