EMAIL THIS PAGE TO A FRIEND
8.43846 EMD Millipore

Methyl caprate

Methyl caprate for synthesis. CAS 110-42-9, molar mass 186.29 g/mol., for synthesis

Synonym: Methyl caprate, Capric acid methyl ester, Decanoic acid methyl ester

  • CAS Number 110-42-9

  • Linear Formula C₁₁H₂₂O₂

  • Molecular Weight 186.29 g/mol

  •  EC Number 203-766-6

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 ml in Glass bottle (8438460025)
bp   >176.7 °C (1013 hPa)
expl. lim.     0.7 %(V)
solubility   0.0038 g/l
density   0.87 g/cm3 (20 °C)
flash point   90 °C
mp   -18 - -10 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Density (d 20 °C/ 4 °C): 0.868 - 0.873
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2915 90 70
Hazard Statement(s) 
H315: Causes skin irritation.
Precautionary Statement(s) 
P262: Do not get in eyes, on skin, or on clothing.
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
RTECS 
HD9830000
Storage class 
10 Combustible liquids not in Storage Class 3
WGK 
WGK 1 slightly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Irritant  Irritant
Categories of danger 
irritant
R Phrase 
R 38
Irritating to skin.
S Phrase 
S 25
Avoid contact with eyes.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?