• USA Home
  • 8.43848 - N-Benzylglycine ethyl ester

EMAIL THIS PAGE TO A FRIEND
8.43848 EMD Millipore

N-Benzylglycine ethyl ester

N-Benzylglycine ethyl ester for synthesis. CAS 6436-90-4, molar mass 193.25 g/mol., for synthesis

  • CAS Number 6436-90-4

  • Linear Formula C₁₁H₁₅NO₂

  • Molecular Weight 193.25 g/mol

  •  EC Number 229-218-6

Purchase

Properties

Related Categories Biochemicals and Reagents More...
density   1.04 g/cm3 (20 °C)
flash point   112 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 96.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.045 - 1.053
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2922 49 85
Storage class 
10 Combustible liquids not in Storage Class 3
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?