• USA Home
  • 8.43850 - 2-Bromomalondialdehyd

EMAIL THIS PAGE TO A FRIEND
8.43850 EMD Millipore

2-Bromomalondialdehyd

2-Bromomalondialdehyd for synthesis. CAS 2065-75-0, molar mass 150.96 g/mol., for synthesis

  • CAS Number 2065-75-0

  • Linear Formula C₃H₃BrO₂

  • Molecular Weight 150.96 g/mol

  •  EC Number 430-470-6

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   5 g in Glass bottle (8438500005)
mp   132 - 136 °C
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 94.0 % (a/a)
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2913 00 00
Hazard Statement(s) 
H302: Harmful if swallowed.
H318: Causes serious eye damage.
Precautionary Statement(s) 
P280: Wear eye protection.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
P313: Get medical advice/ attention.
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS05  GHS05
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Harmful  Harmful
Categories of danger 
harmful, irritant
R Phrase 
R 22-41
Harmful if swallowed.Risk of serious damage to eyes.
S Phrase 
S 26-39
In case of contact with eyes, rinse immediately with plenty of water and seek medical advice.Wear eye/face protection.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?