• USA Home
  • 8.43852 - 1,4-Diamino-2,3-dihydroanthrachinone

EMAIL THIS PAGE TO A FRIEND
8.43852 EMD Millipore

1,4-Diamino-2,3-dihydroanthrachinone

1,4-Diamino-2,3-dihydroanthrachinone for synthesis. CAS 81-63-0, molar mass 240.26 g/mol., for synthesis

  • CAS Number 81-63-0

  • Linear Formula C₁₄H₁₂N₂O₂

  • Molecular Weight 240.26 g/mol

  •  EC Number 201-367-1

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   10 g in Glass bottle (8438520010)
  100 g in Glass bottle (8438520100)
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (HPLC, area%): ≥ 98.0 % (a/a)
  Melting range (lower value): ≥ 253 °C
  Melting range (upper value): ≤ 257 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2922 39 00
RTECS 
CB6570000
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 1 slightly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?