• USA Home
  • 8.43863 - N-Benzyl-N,N-dimethyldodecylammonium chloride

EMAIL THIS PAGE TO A FRIEND
8.43863 EMD Millipore

N-Benzyl-N,N-dimethyldodecylammonium chloride

N-Benzyl-N,N-dimethyldodecylammonium chloride for synthesis. CAS 139-07-1, molar mass 339.99 g/mol., for synthesis

  • CAS Number 139-07-1

  • Linear Formula C₂₁H₃₈N*Cl

  • Molecular Weight 339.99 g/mol

  •  EC Number 205-351-5

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 g in Glass bottle (8438630025)
mp   60 °C
LD 50 oral   LD50 Rat 400 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (argentometric): ≥ 94.0 %
  Identity (IR): passes test
  Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2923 90 00
Hazard Statement(s) 
H302 + H312: Harmful if swallowed or in contact with skin
H314: Causes severe skin burns and eye damage.
H400: Very toxic to aquatic life.
Precautionary Statement(s) 
P273: Avoid release to the environment.
P280: Wear protective gloves/ protective clothing/ eye protection/ face protection.
P301 + P330 + P331: IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
P308 + P310: IF exposed or concerned: immediately call a POISON CENTER or doctor/ physician.
RTECS 
BO6821500
Storage class 
8A Combustible, corrosive hazardous materials
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS05  GHS05
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Pictogram(s) 
GHS09  GHS09
Hazard Symbols 
Corrosive  Corrosive
Hazard Symbols 
Dangerous for the environment  Dangerous for the environment
Declaration (railroad and road) ADR, RID 
UN 1759 , 8, II
Declaration (transport by air) IATA-DGR 
UN 1759 , 8, II
Declaration (transport by sea) IMDG-Code 
UN 1759 , 8, II
Categories of danger 
harmful, corrosive, dangerous for the environment
R Phrase 
R 21/22-34-50
Harmful in contact with skin and if swallowed.Causes burns.Very toxic to aquatic organisms.
S Phrase 
S 26-36/37/39-45-61
In case of contact with eyes, rinse immediately with plenty of water and seek medical advice.Wear suitable protective clothing, gloves and eye/face protection.In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).Avoid release to the environment. Refer to special instructions/ Safety data sheets.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?