• USA Home
  • 8.45029 - Dimethylammonium chloride

EMAIL THIS PAGE TO A FRIEND
8.45029 EMD Millipore

Dimethylammonium chloride

Dimethylammonium chloride Msynth®plus. CAS 506-59-2, chemical formula (CH₃)₂NH * HCl., Msynth®plus

  • CAS Number 506-59-2

  • Linear Formula C₂H₈ClN

  • Molecular Weight 81.54 g/mol

  •  EC Number 208-046-5

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   250 g in Plastic bottle (8450290250)
vapor pressure   <0.1 hPa (25 °C)
mp   160 °C
pH   5 (100 g/l, H₂O, 20 °C)
bulk density   500 kg/m3
LD 50 oral   LD50 Rat 1070 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Colour (visual): white
  Appearance of substance (visual): powder
  Assay (argentometric): ≥ 99.0 %
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2921 11 00
Hazard Statement(s) 
H302: Harmful if swallowed.
Precautionary Statement(s) 
P262: Do not get in eyes, on skin, or on clothing.
RTECS 
IQ0220000
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 1 slightly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Harmful  Harmful
Declaration (railroad and road) ADR, RID 
Kein Gefahrgut
Declaration (transport by air) IATA-DGR 
No Dangerous Good
Declaration (transport by sea) IMDG-Code 
No Dangerous Good
Categories of danger 
harmful
R Phrase 
R 22
Harmful if swallowed.
S Phrase 
S 24/25
Avoid contact with skin and eyes.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?