• USA Home
  • 8.45033 - 1,1-Dichloroethylene (stabilised with hydrochinone monomethyl ether)

EMAIL THIS PAGE TO A FRIEND
8.45033 EMD Millipore

1,1-Dichloroethylene (stabilised with hydrochinone monomethyl ether)

1,1-Dichloroethylene (stabilised with hydrochinone monomethyl ether) Msynth®plus. CAS 75-35-4, chemical formula Cl₂C=CH₂., Msynth®plus

Synonym: Vinylidenchlorid

  • CAS Number 75-35-4

  • Linear Formula C₂H₂Cl₂

  • Molecular Weight 96.94 g/mol

  •  EC Number 200-864-0

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   100 ml in Glass bottle (8450330100)
bp   31.2 - 31.7 °C (1013 hPa)
expl. lim.     8.4 - 16.5 %(V)
vapor pressure   665 hPa (20 °C)
solubility   2.5 g/l
density   1.21 g/cm3 (20 °C)
flash point   -25 °C
autoignition temp.   520 °C
mp   -122 °C
shipped in   ambient
storage temp.   Store at +2°C to +8°C.
specifications   Appearance of substance (visual): clear
  Colour number (Hazen): ≤ 20
  Assay (GC, area%): ≥ 99.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.212 - 1.213
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2903 29 00
Hazard Statement(s) 
H224: Extremely flammable liquid and vapour.
H332: Harmful if inhaled.
H351: Suspected of causing cancer.
Precautionary Statement(s) 
P210: Keep away from heat/sparks/open flames/hot surfaces. No smoking.
P233: Keep container tightly closed.
P240: Ground/bond container and receiving equipment.
P281: Use personal protective equipment as required.
P308 + P313: IF exposed or concerned: Get medical advice/ attention.
P403 + P235: Store in a well-ventilated place. Keep cool.
RTECS 
KV9275000
Storage class 
3 Flammable liquids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS02  GHS02
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Pictogram(s) 
GHS08  GHS08
Hazard Symbols 
Highly Flammable  Highly Flammable
Hazard Symbols 
Harmful  Harmful
Declaration (railroad and road) ADR, RID 
UN 1303 , 3, I
Declaration (transport by air) IATA-DGR 
UN 1303 , 3, I
Declaration (transport by sea) IMDG-Code 
UN 1303 , 3, I, Marine Pollutant: P, Segregation Group: 10 (Liquid halogenated hydrocarbons)
Categories of danger 
extremely flammable, harmful, carcinogenic
R Phrase 
R 12-20-40
Extremely flammable.Harmful by inhalation.Limited evidence of a carcinogenic effect.
S Phrase 
S 7-16-29-36/37-46
Keep container tightly closed.Keep away from sources of ignition - No smoking.Do not empty into drains.Wear suitable protective clothing and gloves.If swallowed, seek medical advice immediately and show this container or label.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?