EMAIL THIS PAGE TO A FRIEND
8.45049 EMD Millipore

Aminopropanol

Aminopropanol (mixture of isomers) Msynth®plus. CAS No. 78-96-6, EC Number 201-162-7., (mixture of isomers) Msynth®plus

Synonym: Aminoisopropyl alcohol, Hydroxypropylamine, Isopropanolamine

  • CAS Number 78-96-6

  • Linear Formula C₃H₉NO

  • Molecular Weight 75.11 g/mol

  •  EC Number 201-162-7

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   100 ml in Glass bottle (8450490100)
bp   160 °C
expl. lim.     1.9 - 10.4 %(V)
vapor pressure   1.93 hPa (20 °C)
density   0.962 g/cm3 (20 °C)
flash point   71 °C
autoignition temp.   335 °C
mp   2 °C
pH   11.3 (10 g/l, H₂O, 20 °C)
LD 50 oral   LD50 Rat 2700 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Colour number (Hazen) (Hazen): ≤ 20
  Assay isomers (GC, area%): ≥ 98.0 % (a/a)
  Density (d 20 °C/ 4 °C): 0.959 - 0.961
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2922 19 00
Hazard Statement(s) 
H312: Harmful in contact with skin.
H314: Causes severe skin burns and eye damage.
Precautionary Statement(s) 
P280: Wear protective gloves/ protective clothing/ eye protection/ face protection.
P301 + P330 + P331: IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
P308 + P310: IF exposed or concerned: immediately call a POISON CENTER or doctor/ physician.
RTECS 
UA5775000
Storage class 
8A Combustible, corrosive hazardous materials
WGK 
WGK 1 slightly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS05  GHS05
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Corrosive  Corrosive
Declaration (railroad and road) ADR, RID 
UN 2735 , 8, II
Declaration (transport by air) IATA-DGR 
UN 2735 , 8, II
Declaration (transport by sea) IMDG-Code 
UN 2735 , 8, II, Segregation Group: 18 (Alkalis)
Categories of danger 
corrosive
R Phrase 
R 34-21
Causes burns.Harmful in contact with skin.
S Phrase 
S 23-26-36-45
Do not breathe vapour.In case of contact with eyes, rinse immediately with plenty of water and seek medical advice.Wear suitable protective clothing.In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?