• USA Home
  • 8.45060 - Trifluoroacetic anhydride

EMAIL THIS PAGE TO A FRIEND
8.45060 EMD Millipore

Trifluoroacetic anhydride

Trifluoroacetic anhydride Msynth®plus. CAS 407-25-0, EC Number 206-982-9, chemical formula (CF₃CO)₂O., Msynth®plus

Synonym: Bis(trifluoracet)-anhydrid, TFAA

  • CAS Number 407-25-0

  • Linear Formula C₄F₆O₃

  • Molecular Weight 210.03 g/mol

  •  EC Number 206-982-9

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   100 ml in Glass bottle (8450600100)
bp   40 °C (1013 hPa)
vapor pressure   405 hPa (20 °C)
density   1.51 g/cm3 (20 °C)
mp   -65 °C
storage temp.   Store below +30°C.
specifications   Colour (visual): colourless
  Appearance of substance (visual): clear
  Assay (GC, area%): ≥ 99.5 % (a/a)
  Density (d 20 °C/ 4 °C): 1.508 - 1.511
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2915 90 70
Hazard Statement(s) 
H314: Causes severe skin burns and eye damage.
H332: Harmful if inhaled.
H335: May cause respiratory irritation.
H412: Harmful to aquatic life with long lasting effects.
EUH014: Reacts violently with water.
Precautionary Statement(s) 
P273: Avoid release to the environment.
P280: Wear protective gloves/ protective clothing/ eye protection/ face protection.
P301 + P330 + P331: IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
P308 + P310: IF exposed or concerned: immediately call a POISON CENTER or doctor/ physician.
P403: Store in a well-ventilated place.
RTECS 
AJ9800000
Storage class 
8B Non-combustible, corrosive hazardous materials
Disposal 
11
Organic acid halides, anhydrides and isocyanates can be added dropwise to an excess of methanol (Cat. No. 822283) to convert them into the corresponding methyl esters or methyl carbamates. If necessary, neutralize with sodium hydroxide solution (Cat. No. 105587). Fill into container A.
Hazard Pictogram(s) 
GHS05  GHS05
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Corrosive  Corrosive
Declaration (railroad and road) ADR, RID 
UN 3265 , 8, I
Declaration (transport by air) IATA-DGR 
UN 3265 , 8, I
Declaration (transport by sea) IMDG-Code 
UN 3265 , 8, I, Segregation Group: 1 (Acids)
Categories of danger 
harmful, corrosive, dangerous for the environment
R Phrase 
R 14-20-35-52/53
Reacts violently with water.Harmful by inhalation.Causes severe burns.Harmful to aquatic organisms, may cause long-term adverse effects in the aquatic environment.
S Phrase 
S 26-36/37/39-45-61
In case of contact with eyes, rinse immediately with plenty of water and seek medical advice.Wear suitable protective clothing, gloves and eye/face protection.In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).Avoid release to the environment. Refer to special instructions/ Safety data sheets.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?