EMAIL THIS PAGE TO A FRIEND
8.45064 EMD Millipore

Trioctylamine

Trioctylamine Msynth®plus. CAS 1116-76-3, molar mass 353.68 g/mol., Msynth®plus

Synonym: N,N-Dioctyl-1-octanamine

  • CAS Number 1116-76-3

  • Linear Formula C₂₄H₅₁N

  • Molecular Weight 353.68 g/mol

  •  EC Number 214-242-1

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   100 ml in Glass bottle (8450640100)
vapor pressure   <0.01 hPa (20 °C)
solubility   <0.0001 g/l
density   0.81 g/cm3 (20 °C)
flash point   168 °C
autoignition temp.   315 °C
mp   -39 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Appearance of substance (visual): liquid
  Colour number (Apha): ≤ 100
  Assay (GC, area%): ≥ 93.0 % (a/a)
  Density (d 20 °C/ 4 °C): 0.808 - 0.813
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2921 19 99
Hazard Statement(s) 
H315: Causes skin irritation.
H319: Causes serious eye irritation.
H411: Toxic to aquatic life with long lasting effects.
Precautionary Statement(s) 
P273: Avoid release to the environment.
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RTECS 
RG8225000
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Pictogram(s) 
GHS09  GHS09
Hazard Symbols 
Irritant  Irritant
Categories of danger 
irritant, dangerous for the environment
R Phrase 
R 36/38-52/53
Irritating to eyes and skin.Harmful to aquatic organisms, may cause long-term adverse effects in the aquatic environment.
S Phrase 
S 61
Avoid release to the environment. Refer to special instructions/ Safety data sheets.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?