• USA Home
  • 8.45065 - Benzyltriethylammonium chloride

EMAIL THIS PAGE TO A FRIEND
8.45065 EMD Millipore

Benzyltriethylammonium chloride

Benzyltriethylammonium chloride Msynth®plus. CAS No. 56-37-1, EC Number 200-270-1., Msynth®plus

Synonym: Triethylbenzylammonium chloride, BTEAC, TEBA

  • CAS Number 56-37-1

  • Linear Formula C₁₃H₂₂ClN

  • Molecular Weight 227.78 g/mol

  •  EC Number 200-270-1

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   100 g in Plastic bottle (8450650100)
solubility   630 g/l
flash point   >275 °C
autoignition temp.   300 °C
mp   190 - 192 °C (decomposition)
pH   6 - 8 (100 g/l, H₂O, 20 °C)
bulk density   700 kg/m3
LD 50 oral   LD50 Rat 2219 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Colour (visual): white to pale yellow
  Appearance of substance (visual): powder
  Assay (argentometric): ≥ 98.0 %
  Identity (IR): passes test
  Acetone: lot specific result

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2923 90 00
Hazard Statement(s) 
H315: Causes skin irritation.
H319: Causes serious eye irritation.
H335: May cause respiratory irritation.
Precautionary Statement(s) 
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RTECS 
BO8275000
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 1 slightly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Irritant  Irritant
Declaration (railroad and road) ADR, RID 
Kein Gefahrgut
Declaration (transport by air) IATA-DGR 
No Dangerous Good
Declaration (transport by sea) IMDG-Code 
No Dangerous Good
Categories of danger 
irritant
R Phrase 
R 36/37/38
Irritating to eyes, respiratory system and skin.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?