• USA Home
  • 8.45068 - Hydroxylamine ( 50% solution in water)

EMAIL THIS PAGE TO A FRIEND
8.45068 EMD Millipore

Hydroxylamine ( 50% solution in water)

Msynth®plus

Purchase

Properties

vapor pressure   14 hPa (20 °C)
density   1.12 g/cm3 (20 °C)
autoignition temp.   215 °C
pH   10.6 (H₂O, 20 °C)
viscosity kinematic   1.435 mm2/s (25 °C)
shipped in   ambient
storage temp.   Store at +15°C to +25°C.
specifications   Colour (visual): colourless
  Appearance of substance (visual): clear
  Assay (manganometric): 48.0 - 52.0 %
  Density (d 20 °C/ 4 °C): 1.120 - 1.123
  Ni (Nickel): lot specific result
  Cr (Chromium): lot specific result
  Cu (Copper): lot specific result
  Mn (Manganese): lot specific result
  Fe (Iron): lot specific result
  Zn (Zinc): lot specific result

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2825 10 00
Hazard Statement(s) 
H290: May be corrosive to metals.
H302: Harmful if swallowed.
H315: Causes skin irritation.
H317: May cause an allergic skin reaction.
H318: Causes serious eye damage.
H335: May cause respiratory irritation.
H351: Suspected of causing cancer.
H373: May cause damage to organs through prolonged or repeated exposure.
H400: Very toxic to aquatic life.
Precautionary Statement(s) 
P273: Avoid release to the environment.
P280: Wear protective gloves.
P280: Wear eye protection.
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P304 + P340: IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
P308 + P313: IF exposed or concerned: Get medical advice/ attention.
Storage class 
8A Combustible, corrosive hazardous materials
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS05  GHS05
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Pictogram(s) 
GHS08  GHS08
Hazard Pictogram(s) 
GHS09  GHS09
Hazard Symbols 
Dangerous for the environment  Dangerous for the environment
Hazard Symbols 
Harmful  Harmful
Declaration (railroad and road) ADR, RID 
UN 2735 , 8, III
Declaration (transport by air) IATA-DGR 
UN 2735 , 8, III
Declaration (transport by sea) IMDG-Code 
UN 2735 , 8, III, Segregation Group: 18 (Alkalis)
Categories of danger 
harmful, irritant, carcinogenic, sensitizing, dangerous for the environment
R Phrase 
R 5-21/22-37/38-40-41-43-48/22-50
Heating may cause an explosion.Harmful in contact with skin and if swallowed.Irritating to respiratory system and skin.Limited evidence of a carcinogenic effect.Risk of serious damage to eyes.May cause sensitisation by skin contact.Harmful: danger of serious damage to health by prolonged exposure if swallowed.Very toxic to aquatic organisms.
S Phrase 
S 26-36/37/39-46-61
In case of contact with eyes, rinse immediately with plenty of water and seek medical advice.Wear suitable protective clothing, gloves and eye/face protection.If swallowed, seek medical advice immediately and show this container or label.Avoid release to the environment. Refer to special instructions/ Safety data sheets.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?