EMAIL THIS PAGE TO A FRIEND
8.45073 EMD Millipore

Glycolic acid

(70% solution in water) Msynth®plus

Synonym: Hydroxyacetic acid

Purchase

Properties

Related Categories Acids, Acids & Bases, Chemical Synthesis, Synthetic Reagents
packaging   100 ml in Plastic bottle (8450730100)
bp   112 °C (1013 hPa)
vapor pressure   27.5 hPa (25 °C)
density   1.26 g/cm3 (20 °C)
mp   10 °C
pH   0.5 (H₂O)
shipped in   ambient
storage temp.   Store at +5°C to +30°C.
specifications   Appearance of substance (visual): clear
  Colour number (Hazen): ≤ 30
  Assay (acidimetric): 69.0 - 74.0 %
  Density (d 20 °C/ 4 °C): 1.260 - 1.280
  Refractive index (n 20°/D): 1.410 - 1.415
  pH-value: 0.0 - 1.0

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2918 19 98
Hazard Statement(s) 
H314: Causes severe skin burns and eye damage.
Precautionary Statement(s) 
P280: Wear protective gloves/ protective clothing/ eye protection/ face protection.
P301 + P330 + P331: IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
P308 + P310: IF exposed or concerned: immediately call a POISON CENTER or doctor/ physician.
Storage class 
8B Non-combustible, corrosive hazardous materials
WGK 
WGK 1 slightly water endangering
Disposal 
4
Liquid organic acids or solutions should be diluted, if necessary, and carefully neutralized with sodium hydrogen carbonate (Cat. No. 106323) or with sodium hydroxide (Cat. No 106462). Before filling into container D, check the pH with pH universal indicator strips (Cat. No. 109535).
Hazard Pictogram(s) 
GHS05  GHS05
Hazard Symbols 
Corrosive  Corrosive
Declaration (railroad and road) ADR, RID 
UN 3265 , 8, II
Declaration (transport by air) IATA-DGR 
UN 3265 , 8, II
Declaration (transport by sea) IMDG-Code 
UN 3265 , 8, II, Segregation Group: 1 (Acids)
Categories of danger 
harmful, corrosive
R Phrase 
R 20-34
Harmful by inhalation.Causes burns.
S Phrase 
S 26-36/37/39-45
In case of contact with eyes, rinse immediately with plenty of water and seek medical advice.Wear suitable protective clothing, gloves and eye/face protection.In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?