• USA Home
  • 8.45091 - Benzamidine hydrochloride hydrate

EMAIL THIS PAGE TO A FRIEND
8.45091 EMD Millipore

Benzamidine hydrochloride hydrate

Benzamidine hydrochloride hydrate Msynth®plus. CAS 206752-36-5, pH 5 (50 g/l, H₂O, 20 °C)., Msynth®plus

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 g in Plastic bottle (8450910025)
mp   80 - 84 °C
pH   5 (50 g/l, H₂O, 20 °C)
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Colour (visual): white to pale yellow
  Appearance of substance (visual): powder
  Assay (argentometric, calc. on anhydrous substance): ≥ 99.0 %
  Water (K. F.): ≤ 14.0 %
  Identity (IR): passes test
  Acetone: lot specific result
  1,4 Dioxane: lot specific result
  Methanol: lot specific result

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2925 29 00
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?