• USA Home
  • 8.45095 - 2-Chlorobenzaldehyde

EMAIL THIS PAGE TO A FRIEND
8.45095 EMD Millipore

2-Chlorobenzaldehyde

2-Chlorobenzaldehyde Msynth®plus. CAS 89-98-5, pH 2.9 (H₂O) (saturated aqueous solution)., Msynth®plus

  • CAS Number 89-98-5

  • Linear Formula C₇H₅ClO

  • Molecular Weight 140.56 g/mol

  •  EC Number 201-956-3

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   250 ml in Glass bottle (8450950250)
vapor pressure   1.7 hPa (50 °C)
solubility   1.8 g/l
density   1.25 g/cm3 (20 °C)
flash point   87 °C
autoignition temp.   397 °C
mp   10 °C
pH   2.9 (H₂O) (saturated aqueous solution)
LD 50 oral   LD50 Rat 2160 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Colour (visual): yellow
  Appearance of substance (visual): clear
  Assay (GC, area%): ≥ 99.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.248 - 1.250
  Free acid: ≤ 1.0 %
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2912 29 00
Hazard Statement(s) 
H314: Causes severe skin burns and eye damage.
Precautionary Statement(s) 
P280: Wear protective gloves/ protective clothing/ eye protection/ face protection.
P301 + P330 + P331: IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
P308 + P310: IF exposed or concerned: immediately call a POISON CENTER or doctor/ physician.
RTECS 
CU5075000
Storage class 
8A Combustible, corrosive hazardous materials
WGK 
WGK 1 slightly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS05  GHS05
Hazard Symbols 
Corrosive  Corrosive
Declaration (railroad and road) ADR, RID 
UN 3265 , 8, III
Declaration (transport by air) IATA-DGR 
UN 3265 , 8, III
Declaration (transport by sea) IMDG-Code 
UN 3265 , 8, III, Segregation Group: 1 (Acids)
Categories of danger 
corrosive
R Phrase 
R 34
Causes burns.
S Phrase 
S 26-45
In case of contact with eyes, rinse immediately with plenty of water and seek medical advice.In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?