• USA Home
  • 8.45114 - 3-Nitro-benzeneboronic acid

EMAIL THIS PAGE TO A FRIEND
8.45114 EMD Millipore

3-Nitro-benzeneboronic acid

3-Nitro-benzeneboronic acid Msynth®plus. CAS 13331-27-6, molar mass 166.93 g/mol., Msynth®plus

  • CAS Number 13331-27-6

  • Linear Formula C₆H₆BNO₄

  • Molecular Weight 166.93 g/mol

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   5 g in Glass bottle (8451140005)
mp   290 - 291 °C (decomposition)
shipped in   ambient
storage temp.   Store below +30°C.

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2931 90 00
RTECS 
CY8980000
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?