EMAIL THIS PAGE TO A FRIEND
8.45130 EMD Millipore

Tropolone

Tropolone Msynth®plus. CAS 533-75-5, molar mass 122.12 g/mol., Msynth®plus

Synonym: 2-Hydroxy-2,4,6-cycloheptatrien-1-one

  • CAS Number 533-75-5

  • Linear Formula C₇H₆O₂

  • Molecular Weight 122.12 g/mol

  •  EC Number 208-577-2

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   5 g in Glass bottle (8451300005)
bp   80 - 84 °C (1.5 hPa)
solubility   40.9 g/l (experimental)
flash point   112 °C
mp   49 - 52 °C
storage temp.   Store at +2°C to +8°C.
specifications   Colour (visual): light yellow to light grey
  Appearance of substance (visual): powder
  Assay (GC, area%): ≥ 99.0 % (a/a)
  Melting range (lower value): ≥ 49 °C
  Melting range (upper value): ≤ 53 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2914 40 90
RTECS 
GU4075000
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Declaration (railroad and road) ADR, RID 
Kein Gefahrgut
Declaration (transport by air) IATA-DGR 
No Dangerous Good
Declaration (transport by sea) IMDG-Code 
No Dangerous Good
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?