• USA Home
  • 8.51002 - 3-Carboxypropanesulfonamide

EMAIL THIS PAGE TO A FRIEND
8.51002 EMD Millipore

3-Carboxypropanesulfonamide

3-Carboxypropanesulfonamide Novabiochem®. CAS 175476-52-5, molar mass 167.18 g/mol., Novabiochem®

Synonym: 4-Sulfamoyl-butyric acid

  • CAS Number 175476-52-5

  • Linear Formula C₄H₉NO₄S

  • Molecular Weight 167.18 g/mol

Purchase

Properties

brand family   Novabiochem
packaging   1 g in Glass bottle (8510020001)
  25 g in Plastic bottle (8510020025)
  5 g in Plastic bottle (8510020005)
mp   90 °C decomposes
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Colour (visual): white to slight yellow to beige
  Appearance of substance (visual): powder
  Assay (NMR): ≥ 90.0 %
  Identity (IR): passes test
  Identity (NMR, 1H): passes test
  Purity (TLC(CMA2)): ≥ 98.0 %
  For TLC-Systems see our General Catalog

Description

Background information

The product number for this product was previously 01-60-0051.

To obtain a certificate of analysis (CoA) of a lot that begins with the letter "A", please select the option in the right hand menu "Request a COA for Lot#s starting with A".

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2935 90 90
Hazard Statement(s) 
H315: Causes skin irritation.
H319: Causes serious eye irritation.
H335: May cause respiratory irritation.
Precautionary Statement(s) 
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Irritant  Irritant
Categories of danger 
irritant
R Phrase 
R 36/37/38
Irritating to eyes, respiratory system and skin.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?