EMAIL THIS PAGE TO A FRIEND
8.51007 EMD Millipore

EDC-HCl

EDC-HCl Novabiochem®. CAS 25952-53-8, chemical formula C₈H₁₇N₃*HCl., Novabiochem®

Synonym: 1-Ethyl-3-(3'-dimethylaminopropyl)carbodiimide · HCl

  • CAS Number 25952-53-8

  • Linear Formula C₈H₁₈ClN₃

  • Molecular Weight 191.70 g/mol

  •  EC Number 247-361-2

Purchase

Properties

brand family   Novabiochem
packaging   25 g in Plastic bottle (8510070025)
mp   110 - 115 °C
shipped in   ambient
storage temp.   Store at +2°C to +8°C.
specifications   Colour (visual): white to slight yellow to beige
  Appearance of substance (visual): powder
  Identity (IR): passes test
  Water (K. F.): ≤ 2.0 %
  Assay (argentometric): ≥ 99.0 %
  Melting range (lower value): ≥ 100 °C
  Melting range (upper value): ≤ 115 °C

Description

Background information

A carbodiimide coupling reagent that generates a urea by-product which can be easily removed from the reaction media by extraction with water [1-3].

The product number for this product was previously 01-62-0011.

To obtain a certificate of analysis (CoA) of a lot that begins with the letter "A", please select the option in the right hand menu "Request a COA for Lot#s starting with A".

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2925 19 95
Hazard Statement(s) 
H315: Causes skin irritation.
H319: Causes serious eye irritation.
H335: May cause respiratory irritation.
Precautionary Statement(s) 
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RTECS 
FF2200000
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Irritant  Irritant
Declaration (railroad and road) ADR, RID 
Kein Gefahrgut
Declaration (transport by air) IATA-DGR 
No Dangerous Good
Declaration (transport by sea) IMDG-Code 
No Dangerous Good
Categories of danger 
irritant
R Phrase 
R 36/37/38
Irritating to eyes, respiratory system and skin.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?