• USA Home
  • 8.52167 - Fmoc-D-Dab(ivDde)-OH

EMAIL THIS PAGE TO A FRIEND
8.52167 EMD Millipore

Fmoc-D-Dab(ivDde)-OH

Fmoc-D-Dab(ivDde)-OH Novabiochem®. CAS 872169-32-9, molar mass 546.65 g/mol., Novabiochem®

Synonym: N-α-Fmoc-N-γ-1-(4,4-dimethyl-2,6-dioxocyclohex-1-ylidene)-3-methylbutyl-D-diaminobutanoic acid

  • CAS Number 872169-32-9

  • Linear Formula C₃₂H₃₈N₂O₆

  • Molecular Weight 546.65 g/mol

Purchase

Properties

Related Categories Amino Acids & Derivatives, Chemical Biology, Chemical Synthesis, Peptide Chemistry
brand family   Novabiochem
packaging   5 g in Plastic bottle (8521670005)
shipped in   ambient
storage temp.   Store at -15°C to -25°C.
specifications   Colour (visual): white to slight yellow to beige
  Appearance of substance (visual): powder
  Identity (IR): passes test
  Enantiomeric purity: ≥ 99.0 % (a/a)
  Purity (TLC(157A)): ≥ 98 %
  Purity (TLC(157B)): ≥ 98 %
  Assay (HPLC, area%): ≥ 95.0 % (a/a)
  Solubility (1 mmole in 2 ml DMF): clearly soluble
  Water (K. F.): ≤ 1.00 %
  For TLC-Systems see our General Catalog

Description

Background information

The product number for this product was previously 04-13-1075.

To obtain a certificate of analysis (CoA) of a lot that begins with the letter "A", please select the option in the right hand menu "Request a COA for Lot#s starting with A".

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2924 29 70
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 2 water endangering
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?