EMAIL THIS PAGE TO A FRIEND
8.53016 EMD Millipore

Boc-Gln(Xan)-OH

Boc-Gln(Xan)-OH Novabiochem®. CAS 55260-24-7, molar mass 426.46 g/mol., Novabiochem®

Synonym: N-α-t.-Boc-γ-xanthyl-L-glutamine

  • CAS Number 55260-24-7

  • Linear Formula C₂₃H₂₆N₂O₆

  • Molecular Weight 426.46 g/mol

Purchase

Properties

Related Categories Amino Acids & Derivatives, Chemical Biology, Chemical Synthesis, Glutamine, Peptide Chemistry More...
brand family   Novabiochem
packaging   100 g in Plastic bottle (8530160100)
  25 g in Plastic bottle (8530160025)
mp   150 °C (decomposition)
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Colour (visual): white to slight yellow to beige
  Appearance of substance (visual): powder
  Identity (IR): passes test
  Enantiomeric purity: ≥ 99.5 % (a/a)
  Purity (TLC(011A)): ≥ 98 %
  Purity (TLC(0811)): ≥ 98 %
  Assay (HPLC, area%): ≥ 98.0 % (a/a)
  Solubility (1 mmole in 2 ml DMF): clear soluble
  Acetate (IC): ≤ 0.1 %
  For TLC-Systems see our General Catalog

Description

Background information

The product number for this product was previously 04-12-0054.

To obtain a certificate of analysis (CoA) of a lot that begins with the letter "A", please select the option in the right hand menu "Request a COA for Lot#s starting with A".

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2932 99 00
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 2 water endangering
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?