• USA Home
  • 8.53104 - Boc-D-Tyr(2-Br-Z)-OH

EMAIL THIS PAGE TO A FRIEND
8.53104 EMD Millipore

Boc-D-Tyr(2-Br-Z)-OH

Boc-D-Tyr(2-Br-Z)-OH Novabiochem®. CAS 81189-61-9, molar mass 494.33 g/mol., Novabiochem®

Synonym: N-α-t.-Boc-O-2-bromobenzyloxycarbonyl-D-tyrosine

  • CAS Number 81189-61-9

  • Linear Formula C₂₂H₂₄BrNO₇

  • Molecular Weight 494.33 g/mol

Purchase

Properties

Related Categories Amino Acids & Derivatives, Chemical Biology, Chemical Synthesis, Peptide Chemistry, Tyrosine More...
brand family   Novabiochem
packaging   5 g in Plastic bottle (8531040005)
mp   90 - 95 °C
shipped in   ambient
storage temp.   Store at -15°C to -25°C.
specifications   Colour (visual): white to slight yellow to beige
  Appearance of substance (visual): powder
  Identity (IR): passes test
  Optical rotation α 25/D (c=1 in methanol): -10.5 - -5.5 °
  Purity (TLC(011A)): ≥ 98 %
  Purity (TLC(0811)): ≥ 98 %
  Solubility (2 mmol in 2 ml CH2Cl2): clearly soluble
  For TLC-Systems see our General Catalog

Description

Background information

The product number for this product was previously 04-13-0050.

To obtain a certificate of analysis (CoA) of a lot that begins with the letter "A", please select the option in the right hand menu "Request a COA for Lot#s starting with A".

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2924 29 70
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 2 water endangering
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?