EMAIL THIS PAGE TO A FRIEND
8.53115 EMD Millipore

Boc-D-Cys(Trt)-OH

Boc-D-Cys(Trt)-OH Novabiochem®. CAS 87494-13-1, molar mass 463.59 g/mol., Novabiochem®

Synonym: N-α-t.-Boc-S-trityl-D-cysteine

  • CAS Number 87494-13-1

  • Linear Formula C₂₇H₂₉NO₄S

  • Molecular Weight 463.59 g/mol

Purchase

Properties

Related Categories Amino Acids & Derivatives, Chemical Biology, Chemical Synthesis, Cysteine/Cystine, Peptide Chemistry More...
brand family   Novabiochem
packaging   25 g in Plastic bottle (8531150025)
shipped in   ambient
storage temp.   Store at -15°C to -25°C.
specifications   Colour (visual): white to slight yellow to beige
  Appearance of substance (visual): powder
  Identity (IR): passes test
  Optical rotation α 25/D (c=1 in ethanol): -28.5 - -23.5 °
  Purity (TLC(0811)): ≥ 98 %
  Solubility (1 mmole in 2 ml DMF): clearly soluble
  For TLC-Systems see our General Catalog

Description

Background information

The product number for this product was previously 04-13-0081.

To obtain a certificate of analysis (CoA) of a lot that begins with the letter "A", please select the option in the right hand menu "Request a COA for Lot#s starting with A".

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2930 90 16
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 2 water endangering
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?