EMAIL THIS PAGE TO A FRIEND
901747 EMD Millipore

Dip tube

for 30 L stainless steel barrel for withdrawal systems with 2" thread adapter

Synonym: Dip tube 30 l barrel for threaded adapter

Purchase

Properties

packaging   1 unit in Plastic bag (9671001041)
shipped in   ambient

Description

Other Notes

To customize the withdrawal systems for every requirement, we provide customers with the appropriate dip tubes for each package size suitable for the various withdrawal systems. Dip tube for withdrawal system 1.06710.0001

Price and Availability

Safety & Documentation

Safety Information

HS Code 
8479 90 70

Documents

Certificate of Analysis

Protocols & Articles

Articles

Safe Handling

Our broad range of withdrawal systems and accessories includes everything you need to ensure safe and easy handling and contamination-free withdrawal of organic inorganics and solvents.
Keywords: Solvents

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?