EMAIL THIS PAGE TO A FRIEND
902888 EMD Millipore

Hood wall duct

with tube connector (OD 10 mm) for G 3/8 internal thread

Purchase

Properties

packaging   1 unit in Plastic bag (9671001069)
shipped in   ambient

Description

Other Notes

Individual solvent supply installations often work with safety cabinets and and fume hoods. This hood wall duct can be used as the link between the inner and outer side of the fume hood. It provides a 10 mm tube end on the outside and a G3/8 inner thread on the inner side.

Price and Availability

Safety & Documentation

Safety Information

HS Code 
7306 50 20
Storage class 
10 - 13 Other liquids and solids

Documents

Certificate of Analysis

Protocols & Articles

Articles

Safety Accessories

Safety Accessories for Bottles Bottles made from glass, aluminum, and HDPE are widely used for packaging and storing solvents and inorganics. We provide a broad range of specially developed safety ac...
Keywords: Solvents

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?