EMAIL THIS PAGE TO A FRIEND
911121 EMD Millipore

Glass bottle 2.5 l

in single UN-approved transport packaging, 1 bottle

Purchase

Properties

packaging   1 unit in Fibre carton (9111212501)
shipped in   ambient

Description

Other Notes

We offer a big variety of packaging materials like empty bottles and containers for your own preparations and mixtures of our reagents.

Price and Availability

Safety & Documentation

Safety Information

HS Code 
7010 90 31
Storage class 
10 - 13 Other liquids and solids

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?