• USA Home
  • 942896 - Stainless steel-coated PTFE-tube (80 cm)

EMAIL THIS PAGE TO A FRIEND
942896 EMD Millipore

Stainless steel-coated PTFE-tube (80 cm)

with rapid-action coupling and pipe connector (O.D.10 mm)

Synonym: Tube VA/PTFE, 80 cm, con-RV10

Purchase

Properties

packaging   1 unit in Plastic Bag(s) (9671009062)
shipped in   ambient

Description

Other Notes

To meet our customers’ individual installation requirements resulting from their specific operating environments, we offer tubings with different endings. The tubing range also includes a stainless steel coated PTFE-tube (length: 80 cm) with coupling (female) Type 21 and pipe connector for 10mm pipes.

Price and Availability

Safety & Documentation

Safety Information

HS Code 
3917 31 00
Storage class 
10 - 13 Other liquids and solids

Documents

Certificate of Analysis

Protocols & Articles

Articles

Safety Accessories

Safety Accessories for Bottles Bottles made from glass, aluminum, and HDPE are widely used for packaging and storing solvents and inorganics. We provide a broad range of specially developed safety ac...
Keywords: Solvents

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?