EMAIL THIS PAGE TO A FRIEND
963533 EMD Millipore

Safety carrier

for2.5 L glass bottles

Purchase

Properties

packaging   1 PC in Fibre carton (9200780001)
shipped in   ambient

Description

Other Notes

This specially developed safety carrier provides an increased protection against glass breakage. The solvent-resistant materials and the high compression strength of the inlay maximize the protection of 4-L-glass-bottles from us during transportation. The high inner volume (20 l) and the leak proof top cover protect against the contact with solvents and vapours in case of glass breakage.

Price and Availability

Safety & Documentation

Safety Information

HS Code 
3923 29 90
Storage class 
10 - 13 Other liquids and solids

Documents

Certificate of Analysis

Protocols & Articles

Articles

Safety Accessories

Safety Accessories for Bottles Bottles made from glass, aluminum, and HDPE are widely used for packaging and storing solvents and inorganics. We provide a broad range of specially developed safety ac...
Keywords: Solvents

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?