EMAIL THIS PAGE TO A FRIEND
01324 Riedel-de Haën

Acetonitrile

TraceSELECT®, for metal speciation analysis, ≥99.9%

Synonym: ACN, Methyl cyanide

  • CAS Number 75-05-8

  • Linear Formula CH3CN

  • Molecular Weight 41.05

  •  Beilstein Registry Number 741857

  •  EC Number 200-835-2

  •  MDL number MFCD00001878

  •  eCl@ss 39031501

Purchase

Properties

Related Categories Acetonitrile and Acetonitrile Solutions, Analytical Reagents, Analytical/Chromatography, Applications, Chromatography Reagents & Solvents,
vapor density   1.41 (vs air)
vapor pressure   72.8 mmHg ( 20 °C)
grade   TraceSELECT®
assay   ≥99.9%
form   liquid
autoignition temp.   973 °F
quality   for metal speciation analysis
shelf life   48 mo.
expl. lim.   16 %
impurities   ≤0.01% water (Karl Fischer)
color   colorless
transmittance   195 nm, ≥80%
  200 nm, ≥95%
  230 nm, 99%
refractive index   n20/D 1.344(lit.)
bp   81-82 °C(lit.)
mp   −48 °C(lit.)
solubility   water: soluble (completely)
density   0.786 g/mL at 25 °C(lit.)
anion traces   bromide (Br-): ≤10 μg/kg
  chloride (Cl-): ≤25 μg/kg
  fluoride (F-): ≤10 μg/kg
  iodide (I-): ≤10 μg/kg
  nitrate (NO3-): ≤25 μg/kg
  nitrite (NO2-): ≤10 μg/kg
  phosphate (PO43-): ≤10 μg/kg
  sulfate (SO42-): ≤10 μg/kg
cation traces   Ag: ≤0.5 μg/kg
  Al: ≤1 μg/kg
  As: ≤1 μg/kg
  Au: ≤0.5 μg/kg
  Ba: ≤0.5 μg/kg
  Be: ≤0.5 μg/kg
  Bi: ≤0.5 μg/kg
  Ca: ≤20 μg/kg
  Cd: ≤0.5 μg/kg
  Ce: ≤0.5 μg/kg
  Co: ≤0.5 μg/kg
  Cr: ≤1 μg/kg
  Cs: ≤0.5 μg/kg
  Cu: ≤1 μg/kg
  Fe: ≤1 μg/kg
  Ga: ≤0.5 μg/kg
  Ge: ≤0.5 μg/kg
  Hg: ≤0.5 μg/kg
  In: ≤0.5 μg/kg
  Ir: ≤0.5 μg/kg
  K: ≤10 μg/kg
  Li: ≤0.5 μg/kg
  Mg: ≤1 μg/kg
  Mn: ≤0.5 μg/kg
  Mo: ≤0.5 μg/kg
  Na: ≤50 μg/kg
  Ni: ≤1 μg/kg
  Pb: ≤0.5 μg/kg
  Pd: ≤0.5 μg/kg
  Pt: ≤0.5 μg/kg
  Rb: ≤0.5 μg/kg
  Rh: ≤0.5 μg/kg
  Ru: ≤0.5 μg/kg
  Sb: ≤0.5 μg/kg
  Se: ≤1 μg/kg
  Sn: ≤0.5 μg/kg
  Sr: ≤0.5 μg/kg
  Ti: ≤1 μg/kg
  Tl: ≤0.5 μg/kg
  V: ≤0.5 μg/kg
  Zn: ≤1 μg/kg
  Zr: ≤0.5 μg/kg

Description

Application

high purity grade for ICP-MS detection

Legal Information

Riedel-de Haën is a registered trademark of Honeywell Specialty Chemicals Seelze GmbH.

TraceSELECT is a registered trademark of Honeywell Specialty Chemicals Seelze GmbH

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 1648 3 / PGII
WGK Germany 
2
RTECS 
AL7700000
Flash Point(F) 
35.6 °F
Flash Point(C) 
2 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?