EMAIL THIS PAGE TO A FRIEND
02877 Riedel-de Haën

Ethanol solution

tested according to Ph.Helv., 70 % (v/v) in H2O

Synonym: Ethanolum 70 per centum1, Ethyl alcohol

  • CAS Number 64-17-5

  • Linear Formula C2H5OH

  • Molecular Weight 46.07

  •  Beilstein Registry Number 1718733

  •  MDL number MFCD00003568

  •  PubChem Substance ID 24845402

Purchase

Properties

Related Categories Analytical/Chromatography, Pharmacopoeia, Pharmacopoeia A-Z, Plastic Bottles, Solvent Bottles,
pharmacopeia   testing conforms to Pharmacopeia
concentration   70 % (v/v) in H2O

Description

Other Notes

Sales restrictions may apply

Legal Information

Riedel-de Haën is a registered trademark of Honeywell Specialty Chemicals Seelze GmbH.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN1170 - class 3 - PG 2 - Ethanol, solution
WGK Germany 
1
Flash Point(F) 
71.6 °F
Flash Point(C) 
22 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?