Synonym: Cristobalite, Quartz, Sand, Silica
CAS Number 60676-86-0
Linear Formula SiO2
Molecular Weight 60.08
EC Number 262-373-8
MDL number MFCD00011232
Popular Documents: RAMAN FTIR (PDF)
| form | powder |
| impurities | ≤0.1% soluble in HCl |
| loss | ≤0.2% loss on ignition |
| color | white |
| refractive index |
n |
| mp | 1610 °C(lit.) |
| density | 2.6 g/mL at 25 °C(lit.) |
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?